Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028852366.1 | XP_028708199.1 | 420 | 130 | ||
XM_028852367.1 | XP_028708200.1 | 420 | 130 | |||
XM_028852368.1 | XP_028708201.1 | 420 | 130 | |||
XM_028852369.1 | XP_028708202.1 | 420 | 130 | |||
XM_028852370.1 | XP_028708203.1 | 420 | 130 | |||
GenBank mRNA | JU326223.1 | - | 153 | 130 | ||
JV614103.1 | - | 327 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |