Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104281.4 | XP_001104281.2 | 727 | 104 | ||
XM_015129863.2 | XP_014985349.2 | 632 | 104 | |||
GenBank mRNA | JU473212.1 | - | 576 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta Inventoried |