Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266123.2 | NP_001253052.1 | 844 | 109 | ||
XM_015110252.2 | XP_014965738.1 | 930 | 109 | |||
GenBank mRNA | JU475026.1 | - | 846 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |