Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194555.3 | NP_001181484.1 | 1443 | 90 | ||
XM_015114180.2 | XP_014969666.1 | 1551 | 90 | |||
XM_015114181.2 | XP_014969667.1 | 1510 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |