Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112077.4 | XP_001112077.1 | 120 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Macaca mulatta |