Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193496.2 | NP_001180425.2 | 753 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |