Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBA57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083460.4 | XP_001083460.1 | 954 | 67 | ||
XM_015114634.2 | XP_014970120.1 | 404 | 67 | |||
XM_015114645.2 | XP_014970131.1 | 896 | 67 | |||
GenBank mRNA | JU325477.1 | - | 246 | 67 | ||
JU482008.1 | - | 490 | 67 | |||
JV644669.1 | - | 507 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |