Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084801.3 | XP_001084801.1 | 340 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |