Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194267.2 | NP_001181196.1 | 591 | 136 | ||
XM_001104266.4 | XP_001104266.1 | 815 | 136 | |||
XM_015112908.2 | XP_014968394.1 | 667 | 136 | |||
GenBank mRNA | JU332441.1 | - | 732 | 136 | ||
JU472973.1 | - | 721 | 136 | |||
JV045208.1 | - | 693 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |