Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260963.1 | NP_001247892.1 | 468 | 86 | ||
XM_028827247.1 | XP_028683080.1 | 764 | 86 | |||
GenBank mRNA | JU337340.1 | - | 574 | 86 | ||
JU473354.1 | - | 440 | 86 | |||
JV045331.1 | - | 468 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |