Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIMAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193516.2 | NP_001180445.2 | 81 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |