Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125076.2 | XP_014980562.2 | 136 | 137 | ||
XM_028839877.1 | XP_028695710.1 | 136 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |