Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261219.1 | NP_001248148.1 | 960 | 67 | ||
GenBank mRNA | JU320362.1 | - | 990 | 67 | ||
JU473345.1 | - | 914 | 67 | |||
JV043871.1 | - | 960 | 67 | |||
JV639083.1 | - | 967 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |