Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261208.1 | NP_001248137.1 | 750 | 120 | ||
XM_015128471.2 | XP_014983957.1 | 892 | 120 | |||
GenBank mRNA | JU337083.1 | - | 771 | 120 | ||
JU477813.1 | - | 747 | 120 | |||
JV048677.1 | - | 750 | 120 | |||
JV665475.1 | - | 756 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |