Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047154.1 | NP_001040619.1 | 645 | 127 | ||
XM_028838494.1 | XP_028694327.1 | 1226 | 127 | |||
XM_028838495.1 | XP_028694328.1 | 826 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |