Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193857.2 | NP_001180786.2 | 1225 | 83 | ||
XM_015151455.2 | XP_015006941.2 | 1005 | 83 | |||
GenBank mRNA | JU326216.1 | - | 796 | 83 | ||
JV648758.1 | - | 806 | 83 |