Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193938.2 | NP_001180867.1 | 88 | 82 | ||
XM_015137938.2 | XP_014993424.1 | 112 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |