Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194736.1 | NP_001181665.1 | 658 | 97 | ||
XM_015118511.2 | XP_014973997.1 | 842 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |