Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194759.3 | NP_001181688.1 | 413 | 94 | ||
XM_028828957.1 | XP_028684790.1 | 396 | 94 | |||
XM_028828958.1 | XP_028684791.1 | 198 | 94 | |||
GenBank mRNA | JV045090.1 | - | 339 | 94 | ||
JV636087.1 | - | 377 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |