Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190863.1 | NP_001177792.1 | 27 | 53 | ||
GenBank mRNA | CB553146.1 | - | 50 | 53 | ||
JU320479.1 | - | 138 | 53 | |||
JU473483.1 | - | 44 | 53 | |||
JV045999.1 | - | 71 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |