Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204311.2 | NP_001191240.2 | 671 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |