Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205183.2 | NP_001192112.1 | 1046 | 97 | ||
XM_028832480.1 | XP_028688313.1 | 1118 | 97 | |||
XM_028832481.1 | XP_028688314.1 | 1778 | 97 | |||
GenBank mRNA | JV051270.1 | - | 168 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |