Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194885.3 | NP_001181814.3 | 2449 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |