Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261095.1 | NP_001248024.1 | 2603 | 107 | ||
XM_015143320.2 | XP_014998806.1 | 3135 | 107 | |||
GenBank mRNA | JU336742.1 | - | 2729 | 107 | ||
JU336743.1 | - | 2603 | 107 | |||
JV560420.1 | - | 2711 | 107 | |||
JV631065.1 | - | 2724 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |