Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261642.1 | NP_001248571.1 | 372 | 74 | ||
XM_015151027.2 | XP_015006513.1 | 443 | 74 | |||
GenBank mRNA | JU473393.1 | - | 437 | 74 | ||
JV046887.1 | - | 372 | 74 | |||
JV528456.1 | - | 433 | 74 | |||
JV614702.1 | - | 443 | 74 |