Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265832.1 | NP_001252761.1 | 1110 | 82 | ||
XM_015132171.2 | XP_014987657.1 | 1308 | 82 | |||
XM_015132176.2 | XP_014987662.1 | 1348 | 82 | |||
XM_015132177.2 | XP_014987663.1 | 1429 | 82 | |||
XM_028843094.1 | XP_028698927.1 | 1342 | 82 | |||
XM_028843095.1 | XP_028698928.1 | 1147 | 82 | |||
XM_028843096.1 | XP_028698929.1 | 1343 | 82 | |||
XM_028843097.1 | XP_028698930.1 | 1343 | 82 | |||
XM_028843098.1 | XP_028698931.1 | 1343 | 82 | |||
XM_028843099.1 | XP_028698932.1 | 1343 | 82 | |||
XM_028843100.1 | XP_028698933.1 | 1343 | 82 | |||
XM_028843101.1 | XP_028698934.1 | 1202 | 82 | |||
XM_028843102.1 | XP_028698935.1 | 1487 | 82 | |||
XM_028843103.1 | XP_028698936.1 | 1429 | 82 | |||
GenBank mRNA | JU323028.1 | - | 1156 | 82 | ||
JU323029.1 | - | 1160 | 82 | |||
JU473220.1 | - | 954 | 82 | |||
JU473221.1 | - | 1131 | 82 | |||
JV565206.1 | - | 1110 | 82 | |||
JV703885.1 | - | 1157 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |