Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265926.1 | NP_001252855.1 | 453 | 98 | ||
GenBank mRNA | JU334895.1 | - | 290 | 98 | ||
JU475901.1 | - | 332 | 98 | |||
JV043553.1 | - | 453 | 98 | |||
JV470922.1 | - | 177 | 98 | |||
JV653353.1 | - | 454 | 98 |