Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266453.1 | NP_001253382.1 | 226 | 59 | ||
XM_028849143.1 | XP_028704976.1 | 247 | 59 | |||
GenBank mRNA | JU334426.1 | - | 225 | 59 | ||
JV045084.1 | - | 226 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |