Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266683.1 | NP_001253612.1 | 449 | 87 | ||
XM_015135190.2 | XP_014990676.1 | 945 | 87 | |||
XM_015135191.2 | XP_014990677.1 | 497 | 87 | |||
GenBank mRNA | JU473731.1 | - | 393 | 87 | ||
JV047825.1 | - | 444 | 87 | |||
JV636301.1 | - | 449 | 87 |