Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278446.1 | NP_001265375.1 | 616 | 81 | ||
GenBank mRNA | JU335774.1 | - | 560 | 81 | ||
JU473363.1 | - | 557 | 81 | |||
JV515827.1 | - | 616 | 81 | |||
JV589729.1 | - | 603 | 81 | |||
JV720426.1 | - | 560 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |