Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd79b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133533.2 | NP_598217.1 | 3 - 4 | 488 | 61 | |
GenBank mRNA | BC088115.1 | - | 3 - 4 | 488 | 7325406 | 61 |
FQ226266.1 | - | 3 - 4 | 485 | 61 | ||
FQ227061.1 | - | 3 - 4 | 483 | 61 | ||
FQ227788.1 | - | 3 - 4 | 477 | 61 | ||
FQ230261.1 | - | 3 - 4 | 479 | 61 | ||
FQ232054.1 | - | 3 - 4 | 479 | 61 | ||
FQ232680.1 | - | 3 - 4 | 479 | 61 | ||
FQ232790.1 | - | 3 - 4 | 502 | 61 | ||
FQ233507.1 | - | 3 - 4 | 477 | 61 | ||
FQ234082.1 | - | 3 - 4 | 462 | 61 | ||
FQ234487.1 | - | 3 - 4 | 477 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |