Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031777.2 | NP_113965.1 | 8 - 9 | 751 | 89 | |
XM_006250266.2 | XP_006250328.1 | 8 - 9 | 936 | 89 | ||
XM_008769765.1 | XP_008767987.1 | 6 - 7 | 587 | 89 | ||
GenBank mRNA | AF026476.1 | - | 6 - 7 | 620 | 89 | |
BC088849.1 | - | 8 - 9 | 751 | 7313699 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |