Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108332.1 | NP_001101802.1 | 6 - 7 | 833 | 98 | |
XM_006249183.2 | XP_006249245.1 | 6 - 7 | 1159 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |