Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007804.1 | NP_001007805.1 | 6 - 7 | 561 | 103 | |
XM_006247185.1 | XP_006247247.1 | 6 - 7 | 694 | 103 | ||
GenBank mRNA | BC086364.1 | - | 6 - 7 | 604 | 7304427 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |