Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024747.1 | NP_001019918.1 | 7 - 8 | 999 | 91 | |
GenBank mRNA | BC098049.1 | - | 7 - 8 | 999 | 7457308 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |