Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108456.3 | NP_001101926.3 | 13 - 14 | 1663 | 68 | |
GenBank mRNA | BC168848.1 | - | 13 - 14 | 1556 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |