Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234528.2 | XP_006234590.1 | 2 - 3 | 1081 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |