Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133320.1 | NP_579854.1 | 1 - 2 | 44 | 63 | |
XM_006246573.2 | XP_006246635.1 | 1 - 2 | 194 | 63 | ||
XM_006246578.2 | XP_006246640.1 | 1 - 2 | 193 | 63 | ||
GenBank mRNA | AY008298.1 | - | 1 - 2 | 44 | 63 | |
BC070909.1 | - | 1 - 2 | 134 | 7103650 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |