Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012106.1 | NP_001012106.1 | 2 - 3 | 231 | 73 | |
XM_006239390.2 | XP_006239452.1 | 4 - 5 | 400 | 73 | ||
XM_006239391.2 | XP_006239453.1 | 2 - 3 | 916 | 73 | ||
XM_008764256.1 | XP_008762478.1 | 3 - 4 | 637 | 73 | ||
GenBank mRNA | BC087687.1 | - | 2 - 3 | 231 | 7307981 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |