Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037784.1 | NP_001032873.1 | 1 - 2 | 197 | 82 | |
XM_006234243.2 | XP_006234305.1 | 1 - 2 | 2412 | 82 | ||
XM_006234244.2 | XP_006234306.1 | 1 - 2 | 2411 | 82 | ||
GenBank mRNA | BC098943.1 | - | 1 - 2 | 197 | 7456980 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |