Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012494.3 | NP_036626.1 | 1 | 758 | 71 | |
XM_006257432.2 | XP_006257494.1 | 3 | 1100 | 71 | ||
XM_008773399.1 | XP_008771621.1 | 3 | 909 | 71 | ||
GenBank mRNA | BC161802.1 | - | 2 | 1003 | 9102491 | 71 |
D16840.1 | - | 3 | 897 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |