Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ren | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012642.4 | NP_036774.4 | 1 - 2 | 113 | 66 | |
GenBank mRNA | BC078878.1 | - | 1 - 2 | 113 | 7126969 | 66 |
J02941.1 | - | 1 - 2 | 306 | 66 | ||
S60054.1 | - | 1 - 2 | 123 | 66 | ||
X07033.1 | - | 1 - 2 | 123 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |