Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013115.1 | NP_037247.1 | 2 - 3 | 935 | 108 | |
XM_006233486.2 | XP_006233548.1 | 2 - 3 | 980 | 108 | ||
XM_006233487.2 | XP_006233549.1 | 2 - 3 | 1023 | 108 | ||
GenBank mRNA | D28581.1 | - | 2 - 3 | 935 | 108 | |
S74898.1 | - | 1 - 2 | 802 | 108 | ||
U26663.2 | - | 1 - 2 | 1159 | 108 | ||
U47287.1 | - | 2 - 3 | 907 | 108 | ||
X83856.1 | - | 2 - 3 | 950 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons |