Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013156.2 | NP_037288.1 | 1 - 2 | 64 | 65 | |
GenBank mRNA | BC063175.1 | - | 1 - 2 | 79 | 6918389 | 65 |
FQ209613.1 | - | 1 - 2 | 75 | 65 | ||
FQ218649.1 | - | 1 - 2 | 73 | 65 | ||
FQ228725.1 | - | 1 - 2 | 75 | 65 | ||
FQ228844.1 | - | 1 - 2 | 75 | 65 | ||
Y00697.1 | - | 1 - 2 | 68 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |