Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013215.1 | NP_037347.1 | 6 - 7 | 891 | 95 | |
GenBank mRNA | AF045464.1 | - | 6 - 7 | 891 | 95 | |
BC078872.1 | - | 6 - 7 | 885 | 7127297 | 95 | |
BC089811.1 | - | 6 - 7 | 854 | 7126881 | 95 | |
X74673.1 | - | 6 - 7 | 888 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |