Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177321.1 | NP_001170792.1 | 3 - 4 | 285 | 73 | |
NM_017200.1 | NP_058896.1 | 2 - 3 | 213 | 73 | ||
XM_006234440.2 | XP_006234502.1 | 3 - 4 | 508 | 73 | ||
XM_006234441.2 | XP_006234503.1 | 3 - 4 | 1529 | 73 | ||
XM_006234444.2 | XP_006234506.1 | 3 - 4 | 1542 | 73 | ||
XM_008761980.1 | XP_008760202.1 | 3 - 4 | 515 | 73 | ||
XM_008761981.1 | XP_008760203.1 | 3 - 4 | 279 | 73 | ||
XM_008761982.1 | XP_008760204.1 | 3 - 4 | 509 | 73 | ||
XM_008761983.1 | XP_008760205.1 | 3 - 4 | 1532 | 73 | ||
XM_008761984.1 | XP_008760206.1 | 3 - 4 | 1545 | 73 | ||
XM_008761985.1 | XP_008760207.1 | 3 - 4 | 521 | 73 | ||
XM_008761986.1 | XP_008760208.1 | 3 - 4 | 515 | 73 | ||
GenBank mRNA | BC087020.1 | - | 3 - 4 | 285 | 7192382 | 73 |
D10926.1 | - | 2 - 3 | 213 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |