Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033956.1 | NP_001029128.1 | 1 - 2 | 98 | 83 | |
NM_017338.2 | NP_059034.1 | 1 - 2 | 98 | 83 | ||
GenBank mRNA | BG663259.1 | - | 1 - 2 | 64 | 83 | |
V01228.1 | - | 1 - 2 | 59 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons |