Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021747.1 | NP_068515.1 | 3 - 4 | 700 | 72 | |
GenBank mRNA | AJ243484.1 | - | 3 - 4 | 700 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |