Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022582.2 | NP_072104.2 | 1 - 2 | 101 | 73 | |
XM_006228668.2 | XP_006228730.1 | 4 - 5 | 359 | 73 | ||
XM_006228670.2 | XP_006228732.1 | 3 - 4 | 531 | 73 | ||
GenBank mRNA | AF036941.1 | - | 1 - 2 | 110 | 73 | |
BI278774.1 | - | 2 - 3 | 73 | |||
U67883.1 | - | 2 - 3 | 110 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |