Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022631.1 | NP_072153.1 | 1 - 2 | 245 | 126 | |
GenBank mRNA | AF188333.1 | - | 1 - 2 | 245 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Rat Wnt Panel Inventoried Probe spans exons |